Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Supplier: VWR International
Certificates
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Environmentally Preferable:
Specifications
- Connection type (port 1):
- Male luer lock
- Male luer lock
- Male luer lock
- Male luer lock
- Male luer lock
- Connection type (port 2):
- ¹/₄-28 UNF
- 10-33
- ¹/₄-18 NPT thread
- ¹/₈-27 NPT thread
- 10-32 special tapered thread
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- Garantía:
- 3 Months