Charging Stands for Picus and eLINE® Pipettes, Sartorius
Catalog # 89171-670
Supplier: SARTORIUS CORPORATION EDI
Specifications
- Descripción:Four-Position Charging Carousel
- Referencia:89171-670
- Nº de proveedor:730991
Specifications
About this item
1-place stand and 4-place carousel for charging and storing Picus or eLINE® pipettes.
The compact design of the 4-pipette charging carousel is ideal for saving bench space in the laboratory. The 4-place rotating head provides easy access to the desired unit.