Orders LinkContactUsLinkComponent
VWR® Macro Pipette Tips
VWR® Macro Pipette Tips
VWR® Macro Pipette Tips
Supplier:  VWR International
Certificates
VWR® Macro Pipette Tips
VWR® Macro Pipette Tips
3 Options Available Below
Product Details & Documents
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Documents
Product Family Options
5 ml macro tips
Product Information
  • Tip Style
    Volume
    Sterile
    Filter
    Length
    Tip Color
    For Use With
    Packed
    Availability
    Price
  • Macro pipette tips
    1000 - 5000 µl
    +
    +
    123.1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Rack of 50 Tips
    Log in for availability
    Log in for availability
    Log in for availability
    Each (50UN)
    Each (50UN) - $1,113.62
    $1,113.62
    Log in for availability
    Case de 10 (50UN)
    Case de 10 (50UN) - $10,393.86
    $10,393.86
    Each (50UN)
       $1,113.62
    Case de 10 (50UN)
       $10,393.86
  • Macro pipette tips
    1000 - 5000 µl
    +
    123.1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Rack of 50 Tips
    Log in for availability
    Log in for availability
    Log in for availability
    Each (50UN)
    Each (50UN) - $288.25
    $288.25
    Log in for availability
    Case de 10 (50UN)
    Case de 10 (50UN) - $2,687.62
    $2,687.62
    Each (50UN)
       $288.25
    Case de 10 (50UN)
       $2,687.62
  • Macro pipette tips
    1000 - 5000 µl
    +
    123.6 mm
    Clear
    Gilson, Rainin Classic and other universal-style pipettors
    Rack of 50 Tips
    Log in for availability
    Log in for availability
    Log in for availability
    Each (50UN)
    Each (50UN) - $555.41
    $555.41
    Log in for availability
    Case de 10 (50UN)
    Case de 10 (50UN) - $5,554.08
    $5,554.08
    Each (50UN)
       $555.41
    Case de 10 (50UN)
       $5,554.08