Orders LinkContactUsLinkComponent
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, VELTEK
Certificates
DECON-PHENE PLUS®, VELTEK
DECON-PHENE PLUS®, VELTEK
2 Options Available Below
Product Details & Documents
DECON-PHENE Plus disinfects controlled areas such as those in compounding pharmacies, biopharmaceutical, pharmaceutical, medical device, industrial facilities, diagnostic manufacturing facilities, and research laboratories. DECON-PHENE Plus is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.
  • Delivered with lot specific analytical, irradiation, and sterility data, tested to current USP compendium
  • Available in our convenient, one-step, ready-to-use, SimpleMix System made with USP Water for Injection (0.25 EU/mL)
  • Multiple convenient container sizes – Unit Dose, 16 oz, 1 Gallon, and 200 L either sterile or non-sterile
Documents
Product Family Options
Información del producto
  • Size
    Form
    Sterility
    Availability
    Price
  • 76628-024
    DECON-PHENE PLUS®
    DECON-PHENE PLUS®
    1 gal.
    Concentrate
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case de 4 (1/Itm/Bx)
    Case de 4 (1/Itm/Bx) - $10,450.00
    $10,450.00
    Case de 4 (1/Itm/Bx)
       $10,450.00
    Supplier Number: DPP-01
  • 76628-010
    DECON-PHENE PLUS®
    DECON-PHENE PLUS®
    1 gal.
    Concentrate
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case de 4 (1/Itm/Bx)
    Case de 4 (1/Itm/Bx) - $15,860.00
    $15,860.00
    Case de 4 (1/Itm/Bx)
       $15,860.00
    Supplier Number: DPP-02