Burette Clamps
Double burette clamps
470105-692
: CLBUR2N
Easily hold two burettes at once.
- PVC coated jaws
- Accepts burettes up to 35 mm O.D.
Aluminum double clamp is designed to securely hold burettes up to 35 mm in diameter. Spring jaws are covered with non-slip PVC sleeves. Compatible with rods up to 13 mm in diameter.
- Description:Double burette clamps