ContactUsLinkComponent
REUZ™ Stainless Steel Beakers with Handle
REUZ™ Stainless Steel Beakers with Handle
Catálogo # 470347-966
Número de proveedor #: UN3003-1000
undefined
REUZ™ Stainless Steel Beakers with Handle
Catálogo # 470347-966
Número de proveedor:  UN3003-1000
Número de proveedor #: UN3003-1000
Iniciar sesión Para disponibilidad
Select an Option
100 ml
Select an Option
100 - 1000 ml
Select an Option
1000 ml
Select an Option
470347-966
Acerca de este artículo
Stainless steel beakers with convenient handles.
  • 304 stainless steel
  • Pouring spout and large handle
  • Easy to clean
  • Impact-resistant
  • Available in 250, 500 and 1000 ml sizes
Manufactured from 304 stainless steel, REUZ™ ('re-use') beakers with handles feature graduations stamped into the inside wall for easy reading of volumes in milliliters or ounces. Include a pouring spout and large, comfortable stainless steel handle. The precision corrosion-resistant stainless construction includes rolled top rims, smooth edges and radiused corners for easy, thorough cleaning. Useful for applications like measuring, mixing, and sample gathering. Can be used with liquids, pastes, powders and granules. Wall thickness is 0.8 mm.
REUZ™ products promote sustainability in all laboratory environments.
Información de pedido: Sold individually.
Frequently Bought Together