Chamber BRITE Autoclave/sterilizer Cleaner
58619-028
- Descripción:Chamber BRITE sterilizer cleaning powder, 10 sachets per box
- Referencia:58619-028
- Nº de proveedor:023212803
Chamber Brite cleaning powder is the easiest way to keep autoclaves clean and free of hard water deposits.
- Simple to use premeasured sachets
- Replaces messy liquid detergents
- Increases sterilization efficiency
- Extends autoclave lifecycle
- Odor free and easy to store
Designed specifically for steam sterilizers, Chamber Brite effectively cleans and descales the reservoir, chamber and tubing in minutes.