Axygen® Automation-Compatible PCR Sealing Mats, Corning
Catalog # 10011-130
Supplier: CORNING CA
Specifications
- Descripción:AxyMat™ Sealing Mat
- Referencia:10011-130
- Nº de proveedor:AM-96-PCR-RD
Specifications
About this item
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
- Minimize evaporation
- Made of research-grade silicone
- Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.