Orders LinkContactUsLinkComponent
KeepBox Attached Lid Containers, Akro-Mils
KeepBox Attached Lid Containers, Akro-Mils
Supplier:   AKRO-MILS
KeepBox Attached Lid Containers, Akro-Mils
Supplier:  AKRO-MILS
Certificates
KeepBox Attached Lid Containers, Akro-Mils
KeepBox Attached Lid Containers, Akro-Mils
2 Options Available Below
Product Details & Documents
KeepBox Attached Lid Containers are the perfect solution for home storage needs and light to medium duty industrial storage.
  • Containers stack when lids are closed and nest when lids are open
  • Textured bottom provides a no-slip grip
Made of durable industrial-grade plastic, KeepBoxes have full-length steel hinge pins for smooth lid operation.
Product Family Options
Información del producto
  • Capacity
    Color
    Dimensions
    Availability
    Price
  • 75855-692
    Attached Lid KeepBox
    Attached Lid KeepBox
    25 lbs.
    Clear/Blue Lid
    15×21.5×12.5" (W×L×H)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (6UN)
    Case (6UN) - $5,863.50
    $5,863.50
    Case (6UN)
       $5,863.50
    Supplier Number: 66486CLDBL
  • 75855-862
    Attached Lid KeepBox
    Attached Lid KeepBox
    75 lbs.
    Clear/Green Lid
    15×21.5×17" (W×L×H)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (3UN)
    Case (3UN) - $4,098.22
    $4,098.22
    Case (3UN)
       $4,098.22
    Supplier Number: 66497CLDGN