PYREX® Funnel, Plain, 60° Angle, Short Stem, Large Size
Short Stem Funnel, 100 mm, 15 mm, 147 mm
Supplier: Corning
Número de proveedor #: 6120-6
PYREX® Funnel, Plain, 60° Angle, Short Stem, Large Size
Catalog # 89090-548
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Short Stem Funnel
- Top I.D.:147 mm
- Stem O.D.:15 mm
- Stem Length:100 mm
- Cat. No.:89090-548
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact