PYREX® Funnels with Long Stem, Corning
Long Stem Funnel, 150 mm, 15 mm, 147 mm
Supplier: CORNING CA
Número de proveedor #: 6100-6
PYREX® Funnels with Long Stem, Corning
Catalog # 89090-546
Supplier: CORNING CA
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.
These funnels are for use in flasks or test tubes without a filter rack. Fluted funnels feature pressed inside flutings to further decrease filtering time.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Long Stem Funnel
- Top I.D.:147 mm
- Stem O.D.:15 mm
- Stem Length:150 mm
- Cat. No.:89090-546
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact