Axygen® Deep Well and Assay Plates, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
- 0.6 ml deep well plate
- ANSI/SLAS 1-2004: Microplates - footprint dimensions
- Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
Especificaciones
- Catalog No:
- 47743-982
- 10011-940
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Especificaciones
Información relacionada
Frequently Bought Together