PYREX® Conical Centrifuge Tubes with White Graduations and Standard Taper Pennyhead Stopper, Corning
Catalog # 89091-440
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volumen:50 ml
- Descripción:Conical Centrifuge Tube with White Graduations
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Top Type:Pennyhead Glass [ST] Stopper, No. 16
- Referencia:89091-440
- O.D.×L:28×151 mm
- Nº de proveedor:8084-50
Specifications
About this item
PYREX®, borosilicate glass.
These tubes have white enamel graduations. The 5 and 15 ml sizes are graduated upward from the tip in 0.1 ml increments. The 50 ml size is graduated upward from tip to 10 ml in 0.5 ml increments, and from 10 ml to 50 in 1 ml increments. The pennyhead stopper is solid.
For stopper only, see Cat. No. 7661.