Corning® Clear Bottom Black and White Polystyrene Microplates, 96 Well, Corning
66025-630
: Corning
Product available on GSA Advantage®
- Esterilidad:Sterile
- Well Volume:360 µl
- Color:Black
- Descripción:Standard plates with lids, Corning CellBIND®
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:RECs, ISO 14001 - Kennebunk
- Referencia:66025-630
- Empaquetado:5/Bag
- Nº de proveedor:3340
PS, flat bottom.
- Bottoms are 60% thinner than conventional PS plates, resulting in lower background fluorescence and enabling readings down to 340 nm
- Opaque walls prevent well-to-well crosstalk
- Optically clear flat bottom profile permits direct microscope viewing
NBS™ = Non Binding Surface.