Falcon® Multiwell Cell Culture Plates, Corning
: Corning
Standard tissue culture (TC) and Corning® Primaria™ surface treatments is available. These plates are provided with various colors to suit various detection methods.
- Crystal-grade virgin polystyrene
- Nonpyrogenic
- Sterilized by gamma irradiation
- Growth area and volume are nominal
Individual and ready-stack (RS) trays are PET (Code 1) and recyclable.
: Convenient, peel-open medical-style packaging for individual product presentation.
Frequently Bought Together