Costar® Multiple Well Cell Culture Plates
Catalog # 470211-268
Supplier: CORNING CA
Specifications
- Well Volume:3.4 ml
- Color:Clear
- Descripción:TC-coated plate (with lid), skirted, wells with tapered wall
- Referencia:470211-268
- Working Volume:0.38 - 0.57 ml
- Well Depth:17 mm
- Bottom Style:Flat bottom
- Nº de proveedor:3526
Specifications
About this item
Clear PS, flat bottom, with lid. Available with a choice of surface treatments.
- Lids with condensation rings to reduce contamination
- Uniform footprint for ease of stacking
- Individual alphanumeric codes for well identification
- Sterilized by gamma radiation
- Certified non-pyrogenic