Corning® 96-Well Solid Black and White Polystyrene Microplates, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimize light scattering.
Especificaciones
- Catalog No:
- 29442-922
- 29444-018
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Especificaciones
Información relacionada
Certificados
Buscar por número de lote Frequently Bought Together