Corning® Gosselin™ Straw Pipettes, Corning
Catalog # 89135-987
Proveedor: CORNING CA
Especificaciones
- Capacidad:1 ml
- Longitud:15.2 cm (6")
- Empacado:100/Bag, 10 Bags/Box
- Referencia:89135-987
- Nº de proveedor:EPI1B-05
Especificaciones
Acerca de este artículo
Straw pipettes made of PP for liquid handling.
- Packed in tamper-evident zip bags
- Sterile (SAL 10-3)
- Universal size
Disposable straw pipettes are ideal for transferring tiny amounts of liquids without the need for an exact volume.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904