ContactUsLinkComponent
CiDecon® II Concentrated Phenolic Liquid Disinfectant, Decon Labs
CiDecon® II Concentrated Phenolic Liquid Disinfectant, Decon Labs
Catálogo # 76417-136
Proveedor:  DECON LABS
undefined
CiDecon® II Concentrated Phenolic Liquid Disinfectant, Decon Labs
Catálogo # 76417-136
Proveedor:  DECON LABS
Número de proveedor:  8514
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
  • Descripción:
    Phenolic disinfectant
  • Tamaño:
    1 gallon
  • Referencia:
    76417-136
Especificaciones
Acerca de este artículo
CiDecon® II phenolic-based disinfectant is a concentrated liquid packaged in 1 gallon bottles.
  • Phenolic-based concentrated disinfecant
  • Dilution is 1 ounce per gallon of water (1:128)
CiDecon® II concentrated phenolic disinfectant is a high pH (alkaline) germicidal cleaner for use on hard surfaces in labs, production areas, on process equipment, or anywhere thorough disinfection efficacy is required.
CiDecon® II can be used for biological decontamination and disinfection. It is effective against a wide range of pathogenic organisms including TB, HIV-1, bacteria, viruses, and fungi. High soil-load and hard water tolerance. It can be alternated with LopHene II if a rotational disinfection program is desired, or where organism resistance is suspected. Conforms to OSHA’s blood borne pathogens standard. EPA registered.