PYREX® Desiccator Lid, Corning
89134-412
: Corning
Product available on GSA Advantage®
- Environmentally Preferable:Waste Reducing
- Referencia:89134-412
- Nº de proveedor:3121-250CO
PYREX® replacement cover for 10.5 L Large Top Desiccator using 24/29 Standard Taper Stopcock.
Stopcock not included.
Height: 109 mm (approx.)
I.D. of Ground Flange: 273.5 mm (approx.)