PYREX® Solution Bottles, Graduated, Corning
With Graduations, 9.5 L (2.5 gal.)
Supplier: Corning
Supplier #: 1596-9L
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- With double scale (liters and gallons) black enamel graduations
- Graduated in 500 ml increments
- Will withstand hot air (dry) or steam (wet) sterilization
- Necks are tooled to increase mechanical strength and for uniform stopper fit
Solution bottles are designed for storage of solutions and are ideal for media preparation.
Documents
Specifications
- Capacity:9.5 L (2.5 gal.)
- Description:With Graduations