Orders LinkContactUsLinkComponent
VeriSpec® Certified Reference Material, KCl Conductivity Standards
VeriSpec® Certified Reference Material, KCl Conductivity Standards
Supplier:   RICCA CHEMICAL CO
VeriSpec® Certified Reference Material, KCl Conductivity Standards
Supplier:  RICCA CHEMICAL CO
Certificates
VeriSpec® Certified Reference Material, KCl Conductivity Standards
VeriSpec® Certified Reference Material, KCl Conductivity Standards
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Colorless Liquid
  • Conductivity at 25 °C: See Ordering Table
Documents
Product Family Options
Información del producto
  • Size
    Conductivity
    Pack type
    Certification
    Availability
    Price
  • 500 ml
    10 µS/cm
    Plastic bottle
    ISO 17025/17034
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,420.49
    $1,420.49
    Each(1/Itm/Bx)
       $1,420.49
    Supplier Number: RV588510-500A
  • 500 ml
    100 µS/cm
    Plastic bottle
    ISO 17025/17034
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,420.49
    $1,420.49
    Each(1/Itm/Bx)
       $1,420.49
    Supplier Number: RV588700-500A
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 