ContactUsLinkComponent
COSA™ CIP 90 Alkaline Cleaner
COSA™ CIP 90 Alkaline Cleaner
Catálogo # 76554-194
undefined
COSA™ CIP 90 Alkaline Cleaner
Catálogo # 76554-194
Número de proveedor:  6301053
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
  • Tamaño:
    5 gallon
  • Referencia:
    76554-194
  • Empaquetado:
    Pail
Especificaciones
Acerca de este artículo
Cosa™ CIP 90 is a mildly alkaline cleaner.
  • Mild alkaline formula allows for dual use in CIP or manual applications
  • Powerful blend of surfactants and sequestrants for broad cleaning ability
  • Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation
For use in CIP pharmaceutical and cosmetic processing equipment, COSA™ CIP 90 is a mildly alkaline, surfactant containing cleaner. The product has broad cleaning ability and is ideal for the removal of organic soils such as fat, oil, and oil-in-water emulsions in piping, mixers, fluid beds, tanks, and storage vessels. The near-neutral formula also allows for manual cleaning.