Orders LinkContactUsLinkComponent
VWR® Lab Tables
VWR® Lab Tables
VWR® Lab Tables
VWR® Lab Tables
A four-leg, flexible lab table system designed for any application.
  • Powder coated steel
  • Chemical resistant epoxy resin
  • Optional shelving
  • Optional casters
  • Accepts suspended cabinets
  • Accepts fixtures (with uprights)
  • 1000 lbs. capacity
This table system includes a number of accessories that can be added very easily, creating your ideal workstation.
 : SEFA tested (SEFA 3 and 10).
 : Work tops and table frames are sold separately. Tables shipped knocked down, and will need assembly. Professional Assembly is highly recommended.
Product Family Options
Light Neutral Table Frames
  • Description
    Capacity
    Worksurface Material
    Base/Leg Material
    Base/Leg Color/Finish
    Height
    W×D
    W×D×H
  • VWR® Lab table frame, no uprights
    1000 lbs. (SEFA)
    Steel, powder coated
    Light neutral
    48×30×37"
    Each (1UN)
    Each (1UN) - $31,666.03
    $31,666.03
    Each (1UN)
       $31,666.03
Black Epoxy Resin Work Tops
  • Description
    Capacity
    Worksurface Material
    Base/Leg Material
    Base/Leg Color/Finish
    Height
    W×D
    W×D×H
  • Kemresin top for VWR® table
    Epoxy resin
    1"
    48×30"
    Each (1UN)
    Each (1UN) - $21,861.91
    $21,861.91
    Each (1UN)
       $21,861.91