Pulsafeeder Seven-Day Timer Pumps
Supplier: EQUIP SOLUTIONS
Certificates
About this item
Reliable metering performance—guided dual check valves provide precise seating and excellent priming and suction lift.
- PVC and PTFE construction with external control
Seven-day timer pumps provide a simple and cost effective metering system for timed applications. Pumps offer eight on/eight off programs per week for maximum adjustability. These pumps feed chemical products on a timed schedule and are commonly used in biocide feed applications. Feed cycle is initiated and controlled by programmable timer.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX74145-10
- ETL regulatory approval:
- Yes
- Duty cycle continuous:
- Yes
- Inlet connection:
- 3/8" OD tubing
- Presión máx.:
- 100 psi
- Viscosidad máx.:
- 1000 cP
- Outlet connection:
- 3/8" OD tubing
- Repetibilidad:
- ±3%
- Suction lift, wet:
- 1219 mm of H₂O (48" H₂O)
- Wetted materials:
- Ceramic, PTFE, PVC
- Garantía:
- 1 year