Orders LinkContactUsLinkComponent
Eldex High-Pressure Piston Pumps
Eldex High-Pressure Piston Pumps
Supplier:   Eldex Corporation
Eldex High-Pressure Piston Pumps
Supplier:  Eldex Corporation
Certificates
Eldex High-Pressure Piston Pumps
1 Options Available Below
Product Details & Documents
Feature micrometer flow adjustment—deliver exact quantities of liquid with minimal pulsation.
  • Withstand up to 5000 psi outlet pressure
  • Dual-piston models have independent piston control—pump a single liquid, or pump two different liquids at a proportional rate
  • Brushless synchronous motors
Inlet accepts 1/8" OD tubing; outlet accepts 1/16" OD high-pressure SS tubing (order separately). Decrease pulsation with pulse dampener model 07115-55.
What's included: All models come with a sturdy aluminum housing and a 7-ft, three-wire cord (115 VAC models have a plug; 220 VAC models have a European plug).
FLUOROLOY G® The Fluorocarbon Co.
KEL-F® 3M Co.
Specifications for Product Family
Especificaciones
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 2500 psi
  • 5000 psi
  • 1000 psi
  • Max. temperature:
  • 120 °C (248 °F)
  • 120 °C (248 °F)
  • 120 °C (248 °F)
  • Power cord:
  • 7-ft, three-wire cord with plug
  • 7-ft, three-wire cord with plug
  • 7-ft, three-wire cord with plug
  • Repeatability:
  • ±0,3%
  • ±0,3%
  • ±0,3%
  • Wetted materials:
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • 316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Flow rate
    Maximum Flow Rate
    Power
    Width
    Length
    Height
    Availability
    Price
  • MFLX07143-70
    High-pressure piston pump, single-piston, 0.1 to 3.0 ml/min, 2500 psi, 115 VAC
    High-pressure piston pump, single-piston, 0.1 to 3.0 ml/min, 2500 psi, 115 VAC
    0.05 ml/stroke
    3 ml/min (60 strokes/min)
    115 VAC; 60 Hz; 0.125 A
    11.4 cm
    19.7 cm
    12.1 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $88,530.00
    $88,530.00
    Each (1UN)
       $88,530.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07143-70
    Duty cycle continuous
    Yes
    Max. pressure
    2500 psi
    Max. temperature
    120 °C (248 °F)
    Power cord
    7-ft, three-wire cord with plug
    Repeatability
    ±0,3%
    Wetted materials
    316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
    Warranty
    1 year
  • MFLX07143-90
    High-pressure piston pump, dual-piston, 0.2 to 10.0 ml/min, 5000 psi, 115 VAC
    High-pressure piston pump, dual-piston, 0.2 to 10.0 ml/min, 5000 psi, 115 VAC
    0.1 ml/stroke
    10 ml/min (100 strokes/min)
    115 VAC; 60 Hz; 1 A
    26.7 cm
    17.8 cm
    15.9 cm
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07143-90
    Duty cycle continuous
    Yes
    Max. pressure
    5000 psi
    Max. temperature
    120 °C (248 °F)
    Power cord
    7-ft, three-wire cord with plug
    Repeatability
    ±0,3%
    Wetted materials
    316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
    Warranty
    1 year
  • MFLX07143-86
    High-pressure piston pump, single-piston, 0.5 to 5.0 ml/min, 1000 psi, 115 VAC
    High-pressure piston pump, single-piston, 0.5 to 5.0 ml/min, 1000 psi, 115 VAC
    0.05 ml/stroke
    6 ml/min (120 strokes/min)
    115 VAC; 60 Hz; 0.125 A
    11.4 cm
    19.7 cm
    12.1 cm
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX07143-86
    Duty cycle continuous
    Yes
    Max. pressure
    1000 psi
    Max. temperature
    120 °C (248 °F)
    Power cord
    7-ft, three-wire cord with plug
    Repeatability
    ±0,3%
    Wetted materials
    316 SS, Fluoroloy G®, gold-plated stainless steel, PCTFE, ruby, sapphire
    Warranty