Hand-Operated Chemical-Resistant Drum Pumps
Supplier: BURKLE INC
Certificates
About this item
Safely transfer aggressive chemicals.
- PTFE and FEP wetted materials withstand harsh chemicals
- Stopcock contains hazardous vapors and prevents dripping
These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).
- Environmentally Preferable:
Specifications
- Outlet connection:
- Spigot
- Discharge tubing
- Spigot
- Discharge tubing
- Tube ext.Ø, inlet:
- 3.2 cm (1.25")
- 3.2 cm (1.25")
- 3.2 cm (1.25")
- 3.2 cm (1.25")
- Tube length, inlet:
- 95 cm (37.4")
- 95 cm (37.4")
- 60 cm (23.622")
- 60 cm (23.622")
- Tube length, outlet:
- 120 cm (47.25")
- 120 cm (47.25")
- Wetted materials:
- PTFE/FEP
- PTFE/FEP
- PTFE/FEP
- PTFE/FEP
- Garantía:
- 15 Months
- 15 Months
- 15 Months
- 15 Months