Orders LinkContactUsLinkComponent
Remote-Control Diaphragm Liquid Dosing Pumps
Remote-Control Diaphragm Liquid Dosing Pumps
Supplier:   KNF NEUBERGER INC
Remote-Control Diaphragm Liquid Dosing Pumps
Supplier:  KNF NEUBERGER INC
Certificates
Remote-Control Diaphragm Liquid Dosing Pumps
2 Options Available Below
Product Details & Documents
Uniformity of discharge ensures low-pulsation transfer process.
  • Quiet, reliable, accurate, and virtually maintenance-free
  • Pumps can run dry continuously without damage
  • Versatile pumps handle flow rates up to 100 ml/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-free dosing.
This pump features a PTFE-covered diaphragm and IP65-rated splash protection, making it resistant to chemical attack and adaptable to fluid characteristics.
Unique drive system ensures maximum uniformity of discharge resulting in a low-pulsation transfer process. Pumps feature either local or external control capabilities including start/stop and reset/prime via TTL contact as well as output signal fault alarm. Models available with a RS-232 port for connectivity.
Delivery information: Includes 5-ft (1.5-m) power cord and two 3-ft (1-m) pieces of FEP inlet/outlet tubing (1/8" OD x 1/16" ID) with UNF 1/4"-28 fittings. Models with RS-232 port also include a cable with D-Sub-9 connector.
Specifications for Product Family
Especificaciones
  • Inlet connection:
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • Max. pressure:
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • 5.86 bar (85 psi)
  • Max. temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • Outlet connection:
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • 5/16" hose barb
  • Power cord:
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • Run dry:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Suction lift, wet:
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • Wetted materials:
  • PVDF head, PTFE diaphragm, FFKM valves
  • Polypropylene head, PTFE diaphragm, FFKM valves
  • PVDF head, PTFE diaphragm, FFKM valves
  • PTFE head, PTFE diaphragm, FFKM valves
  • PVDF head, PTFE diaphragm, FFKM valves
  • Stainless Steel head, PTFE diaphragm, FFKM valves
  • PTFE head, PTFE diaphragm, FFKM valves
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Maximum Flow Rate
    Power
    Height
    Length
    Width
    Availability
    Price
  • MFLX78167-09
    Digital Remote Control Dosing Pump, PVDF head, 1 to 100 ml/min
    Digital Remote Control Dosing Pump, PVDF head, 1 to 100 ml/min
    100 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $56,160.00
    $56,160.00
    Each (1UN)
       $56,160.00
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-09
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    PVDF head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-00
    Digital Control Dosing Metering Pump, PP head, 0.03 to 20 ml/min flow range
    Digital Control Dosing Metering Pump, PP head, 0.03 to 20 ml/min flow range
    20 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Discontinued
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-00
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    Polypropylene head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-01
    Digital Remote Control Dosing Pump, PVDF head, 0.03 to 20 ml/min
    Digital Remote Control Dosing Pump, PVDF head, 0.03 to 20 ml/min
    20 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $60,840.00
    $60,840.00
    Each (1UN)
       $60,840.00
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-01
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    PVDF head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-02
    Digital Control Dosing Metering Pump, PTFE head, 0.03 to 20 ml/min
    Digital Control Dosing Metering Pump, PTFE head, 0.03 to 20 ml/min
    20 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Discontinued
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-02
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    PTFE head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-05
    Digital Remote Control Plus Dosing Pump, PVDF head, 0.03 to 20 ml/min
    Digital Remote Control Plus Dosing Pump, PVDF head, 0.03 to 20 ml/min
    20 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Discontinued
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-05
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    PVDF head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-07
    Digital Remote Control Plus Dosing Pump, SS head, 0.03 to 20 ml/min
    Digital Remote Control Plus Dosing Pump, SS head, 0.03 to 20 ml/min
    20 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Discontinued
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-07
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    Stainless Steel head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
  • MFLX78167-10
    Digital Remote Control Dosing Pump, PTFE head, 1 to 100 ml/min
    Digital Remote Control Dosing Pump, PTFE head, 1 to 100 ml/min
    100 ml/min
    100 - 230 VAC; 50/60 Hz; 0.54 A
    19.1 cm
    15.9 cm
    10.8 cm
    Discontinued
    Especificaciones:
    1 year warranty
    CE Compliant
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX78167-10
    Inlet connection
    5/16" hose barb
    Max. pressure
    5.86 bar (85 psi)
    Max. temperature
    80 °C (176 °F)
    Outlet connection
    5/16" hose barb
    Power cord
    6-ft power cord with 3-prong U.S. plug
    Run dry
    Yes
    Suction lift, wet
    3962 mm of H₂O (156" H₂O)
    Wetted materials
    PTFE head, PTFE diaphragm, FFKM valves
    Warranty
    1 year
    1 year warranty
    CE Compliant
    IP65 washdown
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10
    In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10
    MFLX78167-52
    Log in Para disponibilidad
    Each (1UN) - $3,550.00
Información del producto
  • Catalog #
    Availability
    Price
  • Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 1m, 1 Fitting
    MFLX78167-55
    Log in Para disponibilidad
    Each (1UN) - $2,700.00
Información del producto
  • Catalog #
    Availability
    Price
  • Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
    MFLX78167-56
    Log in Para disponibilidad
    Each (1UN) - $4,760.00
Información del producto
  • Catalog #
    Availability
    Price
  • Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
    Chemicals Valve Kit for Dosing Pumps 78167-00 to -10
    MFLX78167-63
    Log in Para disponibilidad
    Each (1UN) - $13,090.00
Información del producto
  • Catalog #
    Availability
    Price
  • In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
    In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
    MFLX78167-50
    Log in Para disponibilidad
    Each (1UN) - $2,720.00