Pulsafeeder Economical Peristaltic Pumps
Supplier: EQUIP SOLUTIONS
Certificates
About this item
Easy maintenance and no contamination—tubing is the only wetted part.
- Easy tube change-out minimizes downtime between tubing set changes
- Inherent degassing function and rugged gear train offer many years of reliable service
These simple-to-use, self-priming peristaltic pumps deliver worry-free dosing.
Delivery: Includes pump head, two tubing assemblies, and a 6-ft (1.8-m) power cord.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX74130-04
- MFLX74130-07
- ETL regulatory approval:
- Yes
- Yes
- Power cord:
- 6-ft power cord
- 6-ft power cord
- Presión máx.:
- 100 psi
- 80 psi
- Garantía:
- 2 years
- 2 years