Masterflex® Single-Use Bag Port 1 Bottle Cap Thread", Avantor®
Port disc threaded port with 38-400 thread finish engage 8402+c3156 cleanroom manufactured; 10 pack, Port Disc to Thread, Straight, Engage 8402
Catalog # MFLX40331-05
Supplier: VWR International
Masterflex® Single-Use Bag Port 1 Bottle Cap Thread", Avantor®
Catalog # MFLX40331-05
Supplier: VWR International
Jump to:
Product Details & Documents
Large opening makes it suitable for high viscosity liquid and powder applications.
- 38-400 1" Bottle Cap
- 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.
Specifications
- Connection Type:Port Disc to Thread
- Flow pattern:Straight
- Description:Port disc threaded port with 38-400 thread finish engage 8402+c3156 cleanroom manufactured; 10 pack
- Fitting Material:Engage 8402
- Cat. No.:MFLX40331-05
- Regulatory approvals:FDA;USP
- Sterilisation:Gamma irradiation