Orders LinkContactUsLinkComponent
Masterflex® Luer Winged Fitting, Avantor®
Masterflex® Luer Winged Fitting, Avantor®
Masterflex® Luer Winged Fitting, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Luer Winged Fitting, Avantor®
8 Options Available Below
Product Details & Documents
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297
Documents
Specifications for Product Family
Specifications
  • Connection type (port 1):
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • Male Winged Luer
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
Specifications
Product Family Options
Product Information
  • Connection Type
    Flow pattern
    Fitting Material
    Availability
    Price
  • MFLX40325-00
    Male Luer winged 0.120 to 0.150" in crystalVu cleanroom manufactured; 10 pack
    Male Luer winged 0.120 to 0.150" in crystalVu cleanroom manufactured; 10 pack
    Male Luer Winged
    Straight
    CrystalVu
    Log in for availability
    Log in for availability
    Each (10UN)
    Each (10UN) - $223.00
    $223.00
    Each (10UN)
       $223.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-00
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-01
    Male Luer winged 0.120 to 0.150" in polypropylene cleanroom manufactured; 10 pack
    Male Luer winged 0.120 to 0.150" in polypropylene cleanroom manufactured; 10 pack
    Male Luer Winged
    Straight
    PP
    Log in for availability
    Log in for availability
    Each (10UN)
    Each (10UN) - $223.00
    $223.00
    Each (10UN)
       $223.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-01
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-02
    Male Luer winged 0.154 to 0.192" in crystalVu cleanroom manufactured; 10 pack
    Male Luer winged 0.154 to 0.192" in crystalVu cleanroom manufactured; 10 pack
    Male Luer Winged
    Straight
    CrystalVu
    Log in for availability
    Log in for availability
    Each (10UN)
    Each (10UN) - $223.00
    $223.00
    Each (10UN)
       $223.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-02
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-03
    Male Luer winged 0.154 to 0.192" in polypropylene cleanroom manufactured; 10 pack
    Male Luer winged 0.154 to 0.192" in polypropylene cleanroom manufactured; 10 pack
    Male Luer Winged
    Straight
    PP
    Log in for availability
    Log in for availability
    Each (10UN)
    Each (10UN) - $223.00
    $223.00
    Each (10UN)
       $223.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-03
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-04
    Female Luer winged 0.120 to 0.150" in crystalVu - qc; 10 pack
    Female Luer winged 0.120 to 0.150" in crystalVu - qc; 10 pack
    Female Luer Winged
    Straight
    CrystalVu
    Log in for availability
    Log in for availability
    Each
    Each - $607.00
    $607.00
    Each
       $607.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-04
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-05
    Female Luer winged 0.120 to 0.150" in polypropylene - qc; 10 pack
    Female Luer winged 0.120 to 0.150" in polypropylene - qc; 10 pack
    Female Luer Winged
    Straight
    PP
    Log in for availability
    Log in for availability
    Each
    Each - $236.00
    $236.00
    Each
       $236.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-05
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-06
    Female Luer winged 0.154 to 0.192" in crystalVu - qc; 10 pack
    Female Luer winged 0.154 to 0.192" in crystalVu - qc; 10 pack
    Female Luer Winged
    Straight
    CrystalVu
    Log in for availability
    Log in for availability
    Each
    Each - $236.00
    $236.00
    Each
       $236.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-06
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP
  • MFLX40325-07
    Female Luer winged 0.154 to 0.192" in polypropylene - qc; 10 pack
    Female Luer winged 0.154 to 0.192" in polypropylene - qc; 10 pack
    Female Luer Winged
    Straight
    PP
    Log in for availability
    Log in for availability
    Each
    Each - $236.00
    $236.00
    Each
       $236.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX40325-07
    Connection type (port 1)
    Male Winged Luer
    No. of ports
    2
    Regulatory approvals
    FDA;USP