Masterflex® Female Luer Plug Fitting, Avantor®
Acerca de este artículo
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297
6 Options Available Below
Product Details & Documents
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297
Documents
Certificates
Buscar por número de loteSpecifications for Product Family
Especificaciones
- No. of ports:
- 1
- 1
- 1
- 1
- 1
- 1
- Regulatory approvals:
- FDA;USP
- FDA
- FDA;USP
- FDA
- FDA;USP
- FDA;USP
Especificaciones
Product Family Options
Información del producto
- Connection TypeFlow patternFitting MaterialAvailabilityPrice
- MFLX40316-00Female Luer plug in crystalVu cleanroom manufactured; 10 packFemale Luer plug in crystalVu cleanroom manufactured; 10 packFemale Luer CapStraightCrystalVuLog in Para disponibilidadEach (10UN)$231.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-00No. of ports1Regulatory approvalsFDA;USP - MFLX40316-01Female Luer plug in medical nylon cleanroom manufactured; 10 packFemale Luer plug in medical nylon cleanroom manufactured; 10 packFemale Luer CapStraightMedical nylonLog in Para disponibilidadEach (10UN)$186.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-01No. of ports1Regulatory approvalsFDA - MFLX40316-02Female Luer plug in natural PVDF cleanroom manufactured; 10 packFemale Luer plug in natural PVDF cleanroom manufactured; 10 packFemale Luer CapStraightPVDFLog in Para disponibilidadEach (10UN)$431.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-02No. of ports1Regulatory approvalsFDA;USP - MFLX40316-03Female Luer plug in natural nylon cleanroom manufactured; 10 packFemale Luer plug in natural nylon cleanroom manufactured; 10 packFemale Luer CapStraightNylonLog in Para disponibilidadEach (10UN)$186.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-03No. of ports1Regulatory approvalsFDA - MFLX40316-04Female Luer plug in polypropylene cleanroom manufactured; 10 packFemale Luer plug in polypropylene cleanroom manufactured; 10 packFemale Luer CapStraightPPLog in Para disponibilidadEach (10UN)$223.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-04No. of ports1Regulatory approvalsFDA;USP - MFLX40316-05Female Luer plug a tether in polypropylene cleanroom manufactured; 10 packFemale Luer plug a tether in polypropylene cleanroom manufactured; 10 packFemale Luer CapStraightPPLog in Para disponibilidadEach (10UN)$186.00Especificaciones:
More
Specifications
Cat. No.MFLX40316-05No. of ports1Regulatory approvalsFDA;USP