McMillan Flow Flowmeter Accessories
Supplier: MC MILLAN CO
Acerca de este artículo
Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
3 Options Available Below
Product Details & Documents
Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
Specifications for Product Family
Especificaciones
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- AvailabilityPrice
- MFLX32704-52Ryton and Brass Sensors Cable assembly. 30" (0.9 m) LRyton and Brass Sensors Cable assembly. 30" (0.9 m) LLog in Para disponibilidadEach (1UN)$1,050.00Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32704-52Warranty1 yearCE Compliant - MFLX32707-50Power supply/output cable (0 to 5 VDC), 115 VAC.Especificaciones:
More
Specifications
Cat. No.MFLX32707-50Warranty1 year - MFLX32708-50Power supply/input/output cable (0 to 5 VDC), 115 VACPower supply/input/output cable (0 to 5 VDC), 115 VACDiscontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32708-50Warranty1 year - MFLX32709-90Accessory Base plate for RYTON flowmetersEspecificaciones:
More
Specifications
Cat. No.MFLX32709-90Warranty1 year - MFLX32707-55Power supply/output cable (0 to 5 VDC), 230 VACPower supply/output cable (0 to 5 VDC), 230 VACDiscontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32707-55Warranty1 year - Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32708-55Warranty