Reducer Fittings, Compression, Straight
Supplier: MIDWEST VALVE & FITTING INC
About this item
Excellent choice to connect rigid tubing systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
1 Options Available Below
Product Details & Documents
Excellent choice to connect rigid tubing systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
Specifications for Product Family
Specifications
- Catalog No:
- MFLX06381-80
- MFLX06381-81
- MFLX06381-82
- MFLX06381-83
- MFLX31412-25
- MFLX06381-84
- MFLX31107-30
- MFLX31406-23
- MFLX31406-24
- MFLX31406-25
- MFLX31406-26
- MFLX31406-27
- MFLX31412-28
- MFLX31412-54
- MFLX31412-56
- MFLX31412-58
- MFLX30803-60
- Orifice dimensions:
- 0.7925 mm (0.03125")
- 5.555 mm (0.21875")
- 5.555 mm (0.21875")
- 8.73 mm (0.34375")
- 8.73 mm (0.34375")
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Warranty:
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- LTD life time
- 1 Month
- Out of box
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- None
Specifications
Product Family Options
Product Information
- Connection TypeFlow patternFitting MaterialAvailabilityPrice
- MFLX06381-80Compression reducing union, PVDF, ¼" OD × ⅛" ODCompression reducing union, PVDF, ¼" OD × ⅛" ODCompression ReducerStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06381-80Orifice dimensions0.7925 mm (0.03125")SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX06381-81Compression reducing union, PVDF, ⁵⁄₁₆" OD × ¼" ODCompression reducing union, PVDF, ⁵⁄₁₆" OD × ¼" ODCompression ReducerStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06381-81Orifice dimensions5.555 mm (0.21875")SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX06381-82Compression reducing union, PVDF, ⅜" OD × ¼" ODCompression reducing union, PVDF, ⅜" OD × ¼" ODCompression ReducerStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06381-82Orifice dimensions5.555 mm (0.21875")SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX06381-83Compression reducing union, PVDF, ½" OD × ⅜" ODCompression reducing union, PVDF, ½" OD × ⅜" ODCompression ReducerStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06381-83Orifice dimensions8.73 mm (0.34375")SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX31412-25Fitting, Brass, Straight, Compression Reducer, ⅜" OD×¼" ODFitting, Brass, Straight, Compression Reducer, ⅜" OD×¼" ODCompression ReducerStraightBrassLog in for availabilityEach (1UN)$240.00Specifications:
More
Specifications
Cat. No.MFLX31412-25Orifice dimensionsSterilisationWarrantyLTD life time - MFLX06381-84Compression reducing union, PVDF, ⅝" OD × ⅜" ODCompression reducing union, PVDF, ⅝" OD × ⅜" ODCompression ReducerStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06381-84Orifice dimensions8.73 mm (0.34375")SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX31107-30Fitting, Polypropylene, Straight, Compression Reducer, ⅜" OD×¼" ODFitting, Polypropylene, Straight, Compression Reducer, ⅜" OD×¼" ODCompression ReducerStraightPPDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31107-30Orifice dimensionsSterilisationWarrantyOut of box - MFLX31406-23Fitting, 316 Stainless Steel, Straight, Compression Reducer, ¼" OD×⅛" ODFitting, 316 Stainless Steel, Straight, Compression Reducer, ¼" OD×⅛" ODCompression ReducerStraightStainless steel 316Discontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31406-23Orifice dimensionsSterilisationAutoclave, ethylene oxide (ETO), or gammaWarrantyLTD life timeCE certificate - MFLX31406-24Fitting, 316 Stainless Steel, Straight, Compression Reducer, ⁵⁄₁₆" OD×¼" ODFitting, 316 Stainless Steel, Straight, Compression Reducer, ⁵⁄₁₆" OD×¼" ODCompression ReducerStraightStainless steel 316Discontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31406-24Orifice dimensionsSterilisationAutoclave, ethylene oxide (ETO), or gammaWarrantyLTD life timeCE certificate - MFLX31406-25Fitting, 316 Stainless Steel, Straight, Compression Reducer, ⅜" OD×¼" ODFitting, 316 Stainless Steel, Straight, Compression Reducer, ⅜" OD×¼" ODCompression ReducerStraightStainless steel 316Discontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31406-25Orifice dimensionsSterilisationAutoclave, ethylene oxide (ETO), or gammaWarrantyLTD life timeCE certificate - MFLX31406-26Fitting, 316 Stainless Steel, Straight, Compression Reducer, ⅜" OD×⁵⁄₁₆" ODFitting, 316 Stainless Steel, Straight, Compression Reducer, ⅜" OD×⁵⁄₁₆" ODCompression ReducerStraightStainless steel 316Discontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31406-26Orifice dimensionsSterilisationAutoclave, ethylene oxide (ETO), or gammaWarrantyLTD life timeCE certificate - MFLX31406-27Fitting, 316 Stainless Steel, Straight, Compression Reducer, ½" OD×¼" ODFitting, 316 Stainless Steel, Straight, Compression Reducer, ½" OD×¼" ODCompression ReducerStraightStainless steel 316Discontinued-Specifications:CE certificate
More
Specifications
Cat. No.MFLX31406-27Orifice dimensionsSterilisationAutoclave, ethylene oxide (ETO), or gammaWarrantyLTD life timeCE certificate - MFLX31412-28Fitting, Brass, Straight, Compression Reducer, ½" OD×⅜"Fitting, Brass, Straight, Compression Reducer, ½" OD×⅜"Compression ReducerStraightBrassDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31412-28Orifice dimensionsSterilisationWarrantyLTD life time - MFLX31412-54Compression Union, Brass, ⁵⁄₁₆" × ¼", EachCompression Union, Brass, ⁵⁄₁₆" × ¼", EachCompression ReducerStraightBrassDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31412-54Orifice dimensionsSterilisationWarrantyLTD life time - MFLX31412-56Compression Union, Brass, ½" × ¼", EachCompression Union, Brass, ½" × ¼", EachCompression ReducerStraightBrassDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31412-56Orifice dimensionsSterilisationWarrantyLTD life time - MFLX31412-58Compression Union, Brass, ¾" × ½", EachCompression Union, Brass, ¾" × ½", EachCompression ReducerStraightBrassDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX31412-58Orifice dimensionsSterilisationWarrantyLTD life time - MFLX30803-60Fitting, Nylon, Straight, Compression Reducer, ⅜" OD × ¼" ODFitting, Nylon, Straight, Compression Reducer, ⅜" OD × ¼" ODCompression ReducerStraightNylonDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX30803-60Orifice dimensionsSterilisationWarrantyNone