Fittings, Compression Nut Assembly with Ferrule
Supplier: MIDWEST VALVE & FITTING INC
About this item
Can handle pressures up to 225 psi.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
9 Options Available Below
Product Details & Documents
Can handle pressures up to 225 psi.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
Specifications for Product Family
Specifications
- Catalog No:
- MFLX06380-05
- MFLX06380-61
- MFLX06380-65
- MFLX31212-63
- MFLX31212-64
- MFLX31212-65
- MFLX31212-66
- MFLX31212-67
- MFLX31212-68
- MFLX31212-69
- MFLX31212-70
- MFLX31212-71
- MFLX06380-42
- MFLX06380-60
- Sterilisation:
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (ETO)
- Warranty:
- Unknown
- 1 Month
- 1 Month
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Unknown
- 1 Month
Specifications
Product Family Options
Product Information
- Connection TypeFlow patternFitting MaterialAvailabilityPrice
- MFLX06380-05Fitting, Polypropylene, Compression Nut Assembly with Ferrule, ⅛"; 10/PKFitting, Polypropylene, Compression Nut Assembly with Ferrule, ⅛"; 10/PKCompression Nut Assembly with FerruleStraightPPDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06380-05SterilisationEthylene oxide (ETO) onlyWarrantyUnknown - MFLX06380-61Fitting, PVDF, Compression Nut Assembly with Ferrule, ¼" ODFitting, PVDF, Compression Nut Assembly with Ferrule, ¼" ODCompression Nut Assembly with FerruleStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06380-61SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX06380-65Fitting, PVDF, Compression Nut Assembly with Ferrule, ½" ODFitting, PVDF, Compression Nut Assembly with Ferrule, ½" ODCompression Nut Assembly with FerruleStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06380-65SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month - MFLX31212-63Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ½"Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ½"Compression Nut Assembly with FerruleStraightStainless steel 316Log in for availabilityEach (1UN)$305.00Specifications:
More
Specifications
Cat. No.MFLX31212-63SterilisationWarranty1 year - MFLX31212-64Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ⅜"Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ⅜"Compression Nut Assembly with FerruleStraightStainless steel 316Log in for availabilityEach (1UN)$211.00Specifications:
More
Specifications
Cat. No.MFLX31212-64SterilisationWarranty1 year - MFLX31212-65Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ¼"Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ¼"Compression Nut Assembly with FerruleStraightStainless steel 316Log in for availabilityEach (1UN)$168.00Specifications:
More
Specifications
Cat. No.MFLX31212-65SterilisationWarranty1 year - MFLX31212-66Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ⅛"Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ⅛"Compression Nut Assembly with FerruleStraightStainless steel 316Log in for availabilityEach (1UN)$168.00Specifications:
More
Specifications
Cat. No.MFLX31212-66SterilisationWarranty1 year - MFLX31212-67Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ¹⁄₁₆"Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ¹⁄₁₆"Compression Nut Assembly with FerruleStraightStainless steel 316Log in for availabilityEach (1UN)$283.00Specifications:
More
Specifications
Cat. No.MFLX31212-67SterilisationWarranty1 year - MFLX31212-68Fitting, Brass Compression Nut Assembly with Ferrule, ¹⁄₁₆" ODFitting, Brass Compression Nut Assembly with Ferrule, ¹⁄₁₆" ODCompression Nut Assembly with FerruleStraightBrassLog in for availabilityEach (1UN)$89.70Specifications:
More
Specifications
Cat. No.MFLX31212-68SterilisationWarranty1 year - MFLX31212-69Fitting, Brass Compression Nut Assembly with Ferrule, ⅛" ODFitting, Brass Compression Nut Assembly with Ferrule, ⅛" ODCompression Nut Assembly with FerruleStraightBrassLog in for availabilityEach (1UN)$74.10Specifications:
More
Specifications
Cat. No.MFLX31212-69SterilisationWarranty1 year - MFLX31212-70Fitting, Brass Compression Nut Assembly with Ferrule, ¼" ODFitting, Brass Compression Nut Assembly with Ferrule, ¼" ODCompression Nut Assembly with FerruleStraightBrassLog in for availabilityEach (1UN)$58.50Specifications:
More
Specifications
Cat. No.MFLX31212-70SterilisationWarranty1 year - MFLX31212-71Fitting, Brass Compression Nut Assembly with Ferrule, ⅜" ODFitting, Brass Compression Nut Assembly with Ferrule, ⅜" ODCompression Nut Assembly with FerruleStraightBrassLog in for availabilityEach (1UN)$64.40Specifications:
More
Specifications
Cat. No.MFLX31212-71SterilisationWarranty1 year - MFLX06380-42Ferrule nut, polypropylene, ⅝", 10/packFerrule nut, polypropylene, ⅝", 10/packCompression Nut Assembly with FerruleStraightPPDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06380-42SterilisationEthylene oxide (ETO) onlyWarrantyUnknown - MFLX06380-60Fitting, PVDF, Compression Nut Assembly with Ferrule, ⅛" ODFitting, PVDF, Compression Nut Assembly with Ferrule, ⅛" ODCompression Nut Assembly with FerruleStraightPVDFDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX06380-60SterilisationAutoclave or ethylene oxide (ETO)Warranty1 Month