Orders LinkContactUsLinkComponent
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
Product Family Options
  • Description
    Connection Type
    Flow pattern
    Thread size NPT(M) (port 1)
    Thread size NPT(M) (port 2)
    Fitting Material
  • Threaded reducer nipple, PFA, ¼ x ⅛" male NPT
    Threaded Reducer
    Straight
    ¼"
    ⅛"
    PFA
     : -
    CE certificate
    Specifications
    Cat. No.
    MFLX31320-99
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded nipple, PFA, ¼" male NPT Fitting
    Threaded Union
    Straight
    ¼"
    ¼"
    PFA
    Each (1UN)
    Each (1UN) - $594.00
    $594.00
    Each (1UN)
       $594.00
     : PN-4-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-00
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded reducer nipple, PFA, ⅜ x ¼" male NPT
    Threaded Reducer
    Straight
    ⅜"
    ¼"
    PFA
    Each (1UN)
    Each (1UN) - $617.00
    $617.00
    Each (1UN)
       $617.00
     : PNR-64-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-01
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded reducer nipple, PFA, ½ x ¼" male NPT
    Threaded Reducer
    Straight
    ½"
    ¼"
    PFA
    Each (1UN)
    Each (1UN) - $724.00
    $724.00
    Each (1UN)
       $724.00
     : PNR-84-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-03
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded reducer nipple, PFA, ½ x ⅜" male NPT
    Threaded Reducer
    Straight
    ½"
    ⅜"
    PFA
    Each (1UN)
    Each (1UN) - $724.00
    $724.00
    Each (1UN)
       $724.00
     : PNR-86-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-04
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded nipple, PFA, ½" male NPT Fitting
    Threaded Union
    Straight
    ½"
    ½"
    PFA
    Each (1UN)
    Each (1UN) - $738.00
    $738.00
    Each (1UN)
       $738.00
     : PN-8-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-05
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • PN-2-P Threaded nipple, PFA, ⅛" male NPT Fitting
    Threaded Union
    Straight
    ⅛"
    ⅛"
    PFA
     : -
    CE certificate
    Specifications
    Cat. No.
    MFLX31320-98
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded nipple, PFA, ⅜" male NPT Fitting
    Threaded Union
    Straight
    ⅜"
    ⅜"
    PFA
    Each (1UN)
    Each (1UN) - $617.00
    $617.00
    Each (1UN)
       $617.00
     : PN-6-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-02
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded Reducing Nipples, PFA, ¾"to½" NPT(M)
    Threaded Reducer
    Straight
    ¾"
    ½"
    PFA
    Each (1UN)
    Each (1UN) - $822.00
    $822.00
    Each (1UN)
       $822.00
     : PNR-128-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-06
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • Threaded nipple, PFA, ¾" male NPT Fitting
    Threaded Union
    Straight
    ¾"
    ¾"
    PFA
    Each (1UN)
    Each (1UN) - $833.00
    $833.00
    Each (1UN)
       $833.00
     : PN-12-P
    CE certificate
    Specifications
    Cat. No.
    MFLX31321-07
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate