Micropump® B-Mount Gear Pump Heads
Supplier: MICROPUMP INC
Certificates
About this item
Smooth pulseless flow provides precise, leak-free delivery of fluids.
- Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
- Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX74004-03
- MFLX74004-73
- MFLX74004-11
- MFLX74004-75
- MFLX74004-01
- MFLX74004-31
- MFLX74004-33
- MFLX74004-05
- MFLX74004-27
- MFLX74004-29
- MFLX74004-23
- MFLX74004-17
- MFLX74004-71
- MFLX74004-07
- MFLX74004-09
- Material del cuerpo:
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Stainless steel
- Differential pressure:
- 3.5 bar (50 psi)
- 5.2 bar (75 psi)
- 3.5 bar (50 psi)
- 5.5 bar (80 psi)
- 3.5 bar (50 psi)
- 2.8 bar (40 psi)
- 3.5 bar (50 psi)
- 5.2 bar (75 psi)
- 5.2 bar (75 psi)
- 2.8 bar (40 psi)
- 2.8 bar (40 psi)
- 5.2 bar (75 psi)
- 5.2 bar (75 psi)
- 3.5 bar (50 psi)
- 3.5 bar (50 psi)
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Gear material:
- PTFE
- PEEK
- PTFE
- PEEK
- PTFE
- Carbon graphite
- PTFE
- PPS
- PPS
- Carbon graphite
- Carbon graphite
- PPS
- PEEK
- PTFE
- PTFE
- Inlet connection:
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- Internal bypass:
- Yes
- No
- No
- No
- Yes
- No
- Yes
- Yes
- No
- No
- No
- No
- No
- No
- No
- Max. temperature:
- 54 °C (130 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 177 °C (350 °F)
- 54 °C (130 °F)
- 54 °C (130 °F)
- Viscosidad máx.:
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- 1500 cP
- Min. temperature:
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- Min. viscosity:
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- 0.5 cP
- Outlet connection:
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- Particulates:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Seal material:
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- Series:
- GJ
- GA
- GJ
- GJ
- GJ
- GA
- GJ
- GJ
- GA
- GA
- GA
- GJ
- GA
- GJ
- GJ
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year