Value Plastics Luer Fittings and Adapters
Proveedor: VWR International
Certificados
Acerca de este artículo
Premium luer fittings for critical applications.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Manufactured from 100% virgin raw materials in a Class 100,000 environment
- Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX15973-45
- MFLX15973-45BLK
- Connection type (port 1):
- Male Luer with Rotating Lock Ring
- Male Luer with Rotating Lock Ring
- No. of ports:
- 2
- 2
- Garantía:
- None
- None