Flowline Switch-Tek Vibration Level Switch
Vibration Level Switch; 12-36 VDC, 12 - 36 VDC
Catalog # MFLX68869-04
Supplier: FLOWLINE
Número de proveedor #: LZ12-1405
Jump to:
Product Details & Documents
Don’t worry about buildup—automatically adjusts for optimal performance, alerts when cleaning is required.
- Constructed of Glass filled Ryton® and Viton®
SwitchTek vibration level switch is an excellent choice for more challenging application liquids, including those with light to medium coating or scaling-type characteristics. Liquid examples include light sodium hydroxide, brine and copper sulfate solutions.
Especificaciones
- Description:Vibration Level Switch; 12-36 VDC
- Power:12 - 36 VDC
- Cat. No.:MFLX68869-04
- Max. pressure:150 psi
- Min. process temperature:-40 °F
- Relay output:SPST relay rated for 60 VA
- Warranty:1 year
- Max. process temperature:80 °C
- Accuracy of depth range:± 1 mm in water
- Wetted materials:Glass filled Ryton®, Viton®,