ContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Supplier:  PENDOTECH
Certificates
undefined
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Supplier:  PENDOTECH
Certificates
Specifications
  • Precisión:
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • ±2% of reading between 0 and 6 psi, ±3% of reading between 6 and 30 psi, ±5% of reading between 30 and 60 psi
  • Min. temperature:
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • 2 °C
  • Max. temperature:
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • 50 °C
  • Min. pressure:
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • −7 psi
  • -7 psi
  • -7 psi
  • -7 psi
  • Presión máx.:
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • 75 psi
  • Conexión:
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Custom molded water-tight 4 pin connector, signal +/- and excitation +/-
  • Process connection:
  • 1/2" Hose Barb
  • 1/2" Sanitary Flange
  • 1" Hose Barb
  • 1" Sanitary Flange Inlet/Outlet
  • 3/8" Hose Barb
  • 3/4" Hose Barb
  • 1/4" Hose Barb
  • 1/8" Hose Barb
  • 1/2" Hose barb
  • 1/4" Hose Barb
  • 1" Hose Barb
  • 1” Sanitary Flange to 1” Hose Barb
  • 3/8” Hose barb
  • 3/4" Hose Barb
  • 1/2" Hose Barb
  • 3/8" Hose Barb
  • Wetted materials:
  • Polycarbonate
  • Polysulfone
  • Polycarbonate
  • Polysulfone
  • Polysulfone
  • Polycarbonate
  • Polysulfone
  • Polysulfone
  • Polysulfone
  • Polycarbonate
  • Polysulfone
  • Polysulfone
  • Polysulfone
  • Polysulfone
  • Polysulfone
  • Polycarbonate
  • Garantía:
  • 3 Months
  • 2 years
  • 3 Months
  • 2 years
  • 3 Months
  • 3 Months
  • 3 Months
  • 2 years
  • 2 years
  • 3 Months
  • 3 Months
  • 2 years
  • 2 years
  • 3 Months
  • 3 Months
  • 3 Months