Masterflex® Gold-colored Identifier Sanitary Clamps, Avantor®
Supplier: VWR International
Certificates
About this item
Easy visual identification of critical joints for review in your process.
- Constructed of gold-colored 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-colored sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.
- Environmentally Preferable:
Specifications
- Sanitary clamp size (port 1):
- 1-1/2"
- 3/4"
- 2"
- 3"
- 2"
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year