Warren Rupp ATEX Approved Air-Powered Double-Diaphragm Pump, Santoprene; 14 GPM
MFLX74440-70
5 year warranty
- Tasa de flujo máxima:53 L/min (14 gal./min)
- Potencia:Air
- Viscosidad máx.:11000 cP
- Descripción:ATEX Approved Air-Powered Double-Diaphragm Pump, Santoprene; 14 gal./min
- Altura:179.4 mm
- Longitud:25.72 cm
- Referencia:MFLX74440-70
- Anchura:20.96 cm
- Wetted materials:Santoprene®, PTFE
- Conexión de entrada de aire:0.25 NPT(F)
- Duty cycle continuous:Yes
- Inlet connection:0.5 NPT(F)
- Max. air inlet pressure:6.9 bar (100 psi)
- Max. particulate size Ø:3.2 cm (0.125")
- Outlet connection:0.5 NPT(F)
- Run dry:Yes
- Suction lift, wet:3100 mm of H₂O (120" H₂O; 537 mm Hg (8.8" Hg)
- Consumo de aire:27.2 m³/h (16 ft.³/min)
- Max. temperature:66 °C (150 °F)
- Aprobación ATEX:Yes
- Garantía:5 years
Unique configurations to meet your demanding needs.
- High flow rates — up to 106 GPM!
- Air-powered motor eliminates sparking concerns associated with electrical pumps.
- Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing.
ATEX-Approved Nonmetallic Pumps provide 100% groundability with conductive materials of construction and ensuring no static electricity buildup for ignition-free operation. Pumps meet ATEX directives for potentially explosive atmospheres.
- Tasa de flujo máxima:53 L/min (14 gal./min)
- Potencia:Air
- Viscosidad máx.:11000 cP
- Descripción:ATEX Approved Air-Powered Double-Diaphragm Pump, Santoprene; 14 gal./min
- Altura:179.4 mm
- Longitud:25.72 cm
- Environmentally Preferable:
- Referencia:MFLX74440-70
- Anchura:20.96 cm
- Wetted materials:Santoprene®, PTFE
- Conexión de entrada de aire:0.25 NPT(F)
- Duty cycle continuous:Yes
- Inlet connection:0.5 NPT(F)
- Max. air inlet pressure:6.9 bar (100 psi)
- Max. particulate size Ø:3.2 cm (0.125")
- Outlet connection:0.5 NPT(F)
- Run dry:Yes
- Suction lift, wet:3100 mm of H₂O (120" H₂O; 537 mm Hg (8.8" Hg)
- Consumo de aire:27.2 m³/h (16 ft.³/min)
- Max. temperature:66 °C (150 °F)
- Aprobación ATEX:Yes
- Garantía:5 years