Pump Flow Calibration Columns, PVC 40
About this item
Calibration columns provide a simple, economical method for checking pump flow rates.
- Constructed of durable PVC
Columns are graduated in both ml/min and GPH to help improve system accuracy. For maximum convenience and accuracy, choose small capacity columns to check low flow rates; select larger capacity columns for higher flow rates.
To check your flow rate, install column on the suction end of your pump. Turn the pump on, and fill the column to zero (at the top). Using a ball valve, shut off supply from the storage tank and monitor time as fluid is pulled out of the column. At one minute, the fluid level in the column indicates the pump flow rate in ml/min and GPH!
Columns are made of schedule 40 PVC with a double BoPET polyester-film lamination to protect the scale from chemical attack. Permanently attached cap features a threaded vent opening (same size as the inlet port) for venting fluid back into the supply tank.
7 Options Available Below
Product Details & Documents
Calibration columns provide a simple, economical method for checking pump flow rates.
- Constructed of durable PVC
Columns are graduated in both ml/min and GPH to help improve system accuracy. For maximum convenience and accuracy, choose small capacity columns to check low flow rates; select larger capacity columns for higher flow rates.
To check your flow rate, install column on the suction end of your pump. Turn the pump on, and fill the column to zero (at the top). Using a ball valve, shut off supply from the storage tank and monitor time as fluid is pulled out of the column. At one minute, the fluid level in the column indicates the pump flow rate in ml/min and GPH!
Columns are made of schedule 40 PVC with a double BoPET polyester-film lamination to protect the scale from chemical attack. Permanently attached cap features a threaded vent opening (same size as the inlet port) for venting fluid back into the supply tank.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX74600-00
- MFLX74600-01
- MFLX74600-02
- MFLX74600-04
- MFLX74600-05
- MFLX74600-06
- MFLX74600-07
- MFLX74600-08
- Inlet connection:
- 0.5 NPT(F)
- 0.5 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 1 NPT(F)
- 1 NPT(F)
- 2 NPT(F)
- 2 NPT(F)
- Outlet connection:
- 0.5 NPT(F)
- 0.5 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 1 NPT(F)
- 1 NPT(F)
- 2 NPT(F)
- 2 NPT(F)
- Warranty:
- 1 year
- LTD life time
- 1 year
- 1 year
- LTD life time
- 1 year
- 1 year
- 1 year
Specifications
Product Family Options
Product Information
- LengthMaximum Flow RateAvailabilityPrice
- MFLX74600-00Calibration Column, 250 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-00Inlet connection0.5 NPT(F)Outlet connection0.5 NPT(F)Warranty1 year - MFLX74600-01Calibration Column, 100 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-01Inlet connection0.5 NPT(F)Outlet connection0.5 NPT(F)WarrantyLTD life time - MFLX74600-02Calibration Column, 500 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-02Inlet connection0.75 NPT(F)Outlet connection0.75 NPT(F)Warranty1 year - MFLX74600-04Calibration Column, 1000 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-04Inlet connection0.75 NPT(F)Outlet connection0.75 NPT(F)Warranty1 year - MFLX74600-05Calibration Column, 2000 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-05Inlet connection1 NPT(F)Outlet connection1 NPT(F)WarrantyLTD life time - MFLX74600-06Calibration Column, 4000 ml capacitySpecifications:
More
Specifications
Cat. No.MFLX74600-06Inlet connection1 NPT(F)Outlet connection1 NPT(F)Warranty1 year - MFLX74600-07Calibration Column. 10.000 ml capacityCalibration Column. 10.000 ml capacity68.6 cm10000 ml/minLog in for availabilityEach (1UN)$13,970.00Specifications:
More
Specifications
Cat. No.MFLX74600-07Inlet connection2 NPT(F)Outlet connection2 NPT(F)Warranty1 year - MFLX74600-08Calibration Column. 20.000 ml capacityCalibration Column. 20.000 ml capacity116.8 cm20000 ml/minDiscontinued-Specifications:
More
Specifications
Cat. No.MFLX74600-08Inlet connection2 NPT(F)Outlet connection2 NPT(F)Warranty1 year