PendoTECH® Single-Use Pressure Monitoring and Transmitting Systems
Supplier: PENDOTECH
Certificates
About this item
Single-use solution to monitor process pressure.
- Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
- Data output capability to a PC or control system
- User-configurable Min/Max alarm set points
The PressureMAT combines a pressure monitor, data logger, alarm, and transmitter in one simple control box. Designed for use with single-use, in-line pressure sensors. Each channel takes pressure readings once per second. Process pressure is displayed on the easy-to-read LCD. System is easily calibrated to zero through a single button. The PressureMAT may also transmit output via 4-20 mA signal or can provide RS-232 data transfer for use in common PC applications, such as the PressureMAT Data Acquisition Software or Excel®, through WinWedge or Hyper Terminal software.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX19406-09
- MFLX19406-08
- MFLX19406-03
- MFLX19406-02
- MFLX19406-05
- MFLX19406-40
- MFLX19406-04
- MFLX19406-11
- MFLX19406-10
- MFLX19406-07
- MFLX19406-06
- MFLX19406-41
- Precisión:
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- ±0.1% full-scale
- Resolución:
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- 0.1
- Presión máx.:
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- 75 psi
- Unidades de medición:
- bar
- psi
- bar
- psi
- bar
- bar
- psi
- bar
- psi
- bar
- psi
- psi
- Process connection:
- 4 Pressure Inputs
- 4 Pressure Inputs
- 2 Pressure Inputs
- 2 Pressure Inputs
- 3 Pressure Inputs
- 1 Pressure Input
- 3 Pressure Inputs
- 3 Pressure, 1 Flow Input
- 3 Pressure, 1 Flow Inputs
- 4 Pressure Inputs
- 4 Pressure Inputs
- 2 Pressure, 1 Flow Input
- Pantalla:
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- 8-line LCD, backlit
- Salida:
- 4 relays
- 4 relays
- 4 to 20 mA (2) and relays (2); or 4 relays
- 4 to 20 mA (2) and relays (2); or 4 relays
- 4 to 20 mA (3) and relay (1)
- 4 to 20 mA (1) and relay (1)
- 4 to 20 mA (3) and relay (1)
- 4 to 20 mA - 4 outputs
- 4 to 20 mA - 4 outputs
- 4 to 20 mA - 4 outputs
- 4 to 20 mA - 4 outputs
- 4 to 20 mA - 4 outputs
- Power source:
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- VAC
- Tipo de batería:
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- 12 to 24 VDC from 110/220 VAC wall supply provided
- Ancho:
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 11.9 cm (4.75")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- 20.0025 cm (7.875")
- Profundidad:
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- 5.715 cm (2.25")
- Altura:
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.9 cm (4.75")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- 11.43 cm (4.5")
- Garantía:
- 2 years
- 1 year
- 2 years
- 1 year
- 2 years
- 2 years
- 1 year
- 3 years
- 2 years
- 2 years
- 1 year
- 2 years