Orders LinkContactUsLinkComponent
Flowline Switch-Pro Compact Level Controllers and Compact Junction Boxes
 :   FLOWLINE
Flowline Switch-Pro Compact Level Controllers and Compact Junction Boxes
 :  FLOWLINE
Flowline Switch-Pro Compact Level Controllers and Compact Junction Boxes
Flowline Switch-Pro Compact Level Controllers and Compact Junction Boxes
Easily mounts to any sensor or fitting assembly.
  • Rugged Polypropylene enclosure rated NEMA 4X
  • Swivel base enables conduit alignment
  • IP65 rated
The LC10 AC powered controller is commonly used for the automatic filling or emptying of a tank between two sensors. A single latched relay performs the automatic control with valves or pumps rated up to ½ horsepower. Combine the LC11 controller with Smart Trak and two level switches to complete your compact lift station package. The general purpose, NEMA 4X controller mounts on any sensor or fitting assembly. The optional flash alarm brings immediate attention to process conditions. Package the controller with any of our level switches.
The LC06-1001 junction box is commonly used for the integral wiring termination of a single level or flow switch. The rugged NEMA 4X enclosure includes a 300-degree swivel base for fast and easy conduit alignment.
  • Enclosure material:
  • Polypropylene
  • Min. process temperature:
  • -40 °F
  • -40 °F
  • -40 °F
  • -40 °F
  • Max. process temperature:
  • 70 °C
  • 70 °C
  • 70 °C
  • 70 °C
  • Power consumption:
  • 5 W
  • 5 W
  • 5 W
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • Description
  • Single Sensor Compact Level Controller, 3/4" NPT
    Each (1UN)
    Each (1UN) - $8,700.00
    $8,700.00
    Each (1UN)
       $8,700.00
    IP65 washdown
    Specifications
    Cat. No.
    MFLX43300-22
    Enclosure material
    Min. process temperature
    -40 °F
    Max. process temperature
    70 °C
    Power consumption
    5 W
    Warranty
    1 year
    IP65 washdown
  • Dual Sensor Compact Level Controller, 3/4" NPT
    IP65 washdown
    Specifications
    Cat. No.
    MFLX43300-24
    Enclosure material
    Min. process temperature
    -40 °F
    Max. process temperature
    70 °C
    Power consumption
    5 W
    Warranty
    1 year
    IP65 washdown
  • Compact Level Switch Junction Box, 6 Pole Terminal Strip, 1-2 Sensors, 3/4" NPT
    IP65 washdown
    Specifications
    Cat. No.
    MFLX43300-41
    Enclosure material
    Polypropylene
    Min. process temperature
    -40 °F
    Max. process temperature
    70 °C
    Power consumption
    Warranty
    1 year
    IP65 washdown
  • Strobe Alert Single Sensor Controller, 3/4" NPT
    Each (1UN)
    Each (1UN) - $10,710.00
    $10,710.00
    Each (1UN)
       $10,710.00
    Specifications
    Cat. No.
    MFLX43300-43
    Enclosure material
    Min. process temperature
    -40 °F
    Max. process temperature
    70 °C
    Power consumption
    5 W
    Warranty
    1 year