Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 1 Port, 3" Tri-Clamp; 1/EA
Catalog # MFLX30542-20
Supplier: VWR International
Specifications
- Sanitary clamp size (Port 1):3"
- Descripción:Sanitary Sampling Gasket, Silicone, 1 Port, 3" Tri-Clamp; 1/EA
- Garantía:1 year
- Referencia:MFLX30542-20
- Fitting Material:Platinum-Cured Silicone
- Sanitary Gasket Size:3"
Specifications
About this item
Easily sample fluid in critical high-purity system.
- One internal port accepts thermocouple probe
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.
- Sanitary clamp size (Port 1):3"
- Descripción:Sanitary Sampling Gasket, Silicone, 1 Port, 3" Tri-Clamp; 1/EA
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX30542-20
- Fitting Material:Platinum-Cured Silicone
- Sanitary Gasket Size:3"