Orders LinkContactUsLinkComponent
PFA Compression Female Threaded Adapter Elbows
PFA Compression Female Threaded Adapter Elbows
PFA Compression Female Threaded Adapter Elbows
PFA Compression Female Threaded Adapter Elbows
Create a secure connection with compression fittings.
  • Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 18 months
  • 18 months
Product Family Options
  • Description
    Connection Type
    Flow pattern
    Thread size NPT(F) (port 2)
    Fitting Material
  • Compression Female Threaded Adapter Elbow, PFA, ⅜" OD X ⅜" NPT(F)
    Compression to Thread
    Elbow
    ⅜"
    PFA
    Each (1UN)
    Each (1UN) - $1,715.00
    $1,715.00
    Each (1UN)
       $1,715.00
    Specifications
    Cat. No.
    MFLX31321-83
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • Compression Thread Adapter, Elbow, PFA, ½" OD×½" NPT(F)
    Compression to Thread
    Elbow
    ½"
    PFA
    Specifications
    Cat. No.
    MFLX31321-84
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months