PFA Compression Female Threaded Adapter Elbows
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
- Catalog No:
- MFLX31321-83
- MFLX31321-84
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Warranty:
- 18 months
- 18 months
Product Family Options
- DescriptionConnection TypeFlow patternThread size NPT(F) (port 2)Fitting Material
- Compression Female Threaded Adapter Elbow, PFA, ⅜" OD X ⅜" NPT(F)Compression to ThreadElbow⅜"PFAEach (1UN)$1,715.00
Specifications
Cat. No.MFLX31321-83SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - Compression Thread Adapter, Elbow, PFA, ½" OD×½" NPT(F)Compression to ThreadElbow½"PFA-
Specifications
Cat. No.MFLX31321-84SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months