Orders LinkContactUsLinkComponent
Suction Line Pump Filter Strainers
Suction Line Pump Filter Strainers
Supplier:   SPECIALTY MFG CO
Suction Line Pump Filter Strainers
Supplier:  SPECIALTY MFG CO
Certificates
Suction Line Pump Filter Strainers
19 Options Available Below
Product Details & Documents
Protect pump from sediments filtering into suction line.
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
Specifications for Product Family
Especificaciones
  • Outlet connection:
  • 0.25 NPT(F)
  • 0.25 NPT(F)
  • 0.375 NPT(F)
  • 0.375 NPT(F)
  • 0.375 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1.5 NPT(F)
  • 1.5 NPT(F)
  • 1.5 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • Wetted materials:
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • Nylon and Stainless Steel
  • PVC and Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 6 months
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 6 months
  • 1 year
  • 1 year
  • 6 months
  • 1 year
  • 1 year
  • 6 months
Especificaciones
Product Family Options
Información del producto
  • Inlet Connection
    Availability
    Price
  • MFLX74604-02
    Suction Line Strainer, PVC, 1/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/4" NPT(F), 40 Mesh (stainless steel)
    0.25 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $213.00
    $213.00
    Each (1UN)
       $213.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-02
    Outlet connection
    0.25 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-04
    Suction Line Strainer, PVC, 1/4" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/4" NPT(F), 80 Mesh (stainless steel)
    0.25 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $213.00
    $213.00
    Each (1UN)
       $213.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-04
    Outlet connection
    0.25 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-10
    Suction Line Strainer, PVC, 3/8" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 20 Mesh (stainless steel)
    0.375 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-10
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-12
    Suction Line Strainer, PVC, 3/8" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 40 Mesh (stainless steel)
    0.375 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-12
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-14
    Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
    0.375 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-14
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-20
    Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
    0.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-20
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-22
    Suction Line Strainer, PVC, 1/2" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 40 Mesh (stainless steel)
    0.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-22
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-23
    Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel)
    0.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-23
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 months
  • MFLX74604-24
    Suction Line Strainer, PVC, 1/2" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 80 Mesh (stainless steel)
    0.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-24
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-30
    Suction Line Strainer, PVC, 3/4" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 20 Mesh (stainless steel)
    0.75 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $281.00
    $281.00
    Each (1UN)
       $281.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-30
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-32
    Suction Line Strainer, PVC, 3/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 40 Mesh (stainless steel)
    0.75 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $281.00
    $281.00
    Each (1UN)
       $281.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-32
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-34
    Suction Line Strainer, PVC, 3/4" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 80 Mesh (stainless steel)
    0.75 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $293.00
    $293.00
    Each (1UN)
       $293.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-34
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-42
    Suction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
    1 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-42
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-43
    Suction Line Strainer, PVC, 1" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 50 Mesh (stainless steel)
    1 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-43
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 months
  • MFLX74604-44
    Suction Line Strainer, PVC, 1" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 80 Mesh (stainless steel)
    1 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-44
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-52
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 40 Mesh stainless steel
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 40 Mesh stainless steel
    1.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $504.00
    $504.00
    Each (1UN)
       $504.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-52
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-53
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 50 Mesh (stainless steel)
    1.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $533.00
    $533.00
    Each (1UN)
       $533.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-53
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 months
  • MFLX74604-54
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 80 Mesh (stainless steel)
    1.5 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $533.00
    $533.00
    Each (1UN)
       $533.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-54
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74605-12
    Suction Line Strainer, Nylon, 3/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, Nylon, 3/4" NPT(F), 40 Mesh (stainless steel)
    0.75 NPT(F)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $701.00
    $701.00
    Each (1UN)
       $701.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74605-12
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    Nylon and Stainless Steel
    Warranty
    1 year
  • MFLX74604-33
    Suction Line Strainer, PVC, 3/4" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 50 Mesh (stainless steel)
    0.75 NPT(F)
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX74604-33
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 months