Warren Rupp Air-Operated Double Diaphragm Pumps
Supplier: WARREN RUPP
Certificates
About this item
Complies for applications in food processing, pharmaceutical, and cosmetic applications.
- Handle abrasive and shear-sensitive materials
- Self-priming up to 24 ft of water
- Deadhead against closed discharge without damage
These Warren Rupp Air-Operated Double Diaphragm Pumps are available in two compliance models. The USDA certified pump is ideal for clean-in-place sanitary installations. The FDA-compliant pumps are ideal for food processing, pharmaceutical, and cosmetic applications.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX74440-50
- Consumo de aire:
- 76.5 m³/h (45 ft.³/min)
- Conexión de entrada de aire:
- 0.75 NPT(F)
- Duty cycle continuous:
- Yes
- Inlet connection:
- 1-1/2" Tri-Clamp®
- Max. air inlet pressure:
- 6.9 bar (100 psi)
- Min. temperature:
- –23 °C (–10 °F)
- Max. temperature:
- 88 °C (190 °F)
- Viscosidad máx.:
- 11000 cP
- Outlet connection:
- 1-1/2" Tri-Clamp®
- Run dry:
- Yes
- Suction lift, wet:
- 7300 mm of H₂O (288" H₂O; 227 mm Hg (21" Hg)
- Wetted materials:
- 316 Stainless Steel, Santoprene®
- Garantía:
- 5 years