24 VDC Motor Polypropylene Standard Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
Certificates
About this item
DC-Powered motors are lightweight and portable – excellent for outdoor sampling and transfer of fluids.
- Sealless pump is great when used with chemically aggressive water samples — no replacement of dirty seals
- Compact, brushless design
- Internal cooling circuit reduces bearing temperature
- Pinpoint bearings allow for run dry capability up to 8 hours
These magnetic drive pumps feature continuous duty 24 VDC motors. Pumps are ideal for use with chemically aggressive water samples as they are constructed of glass fiber-reinforced polypropylene front and rear casings, Viton® o-ring, and either PPS or PTFE bearings.
Delivery: Includes 6 ft (1.8 m) cord with stripped ends.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX72008-20
- Duty cycle continuous:
- Yes
- Inlet connection:
- 0.75 NPT(F)
- Presión máx.:
- 1.2 bar (16.9 psi)
- Max. temperature:
- 40 °C (104 °F)
- Outlet connection:
- 0.75 NPT(F)
- Run dry:
- Yes
- Garantía:
- 1 year